Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012721.2 | NP_036853.2 | 11 | 1205 | ||
XM_008768851.1 | XP_008767073.1 | 10 | 1476 | |||
XM_008768852.1 | XP_008767074.1 | 10 | 1161 | |||
GenBank mRNA | CO554543.1 | - | 1 | 56 | ||
X92070.1 | - | 11 | 1086 | |||
X97376.1 | - | 11 | 1216 |