Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012735.2 | NP_036867.1 | 1 | 213 | ||
GenBank mRNA | AF027179.1 | - | 1 | 17 | ||
M68971.1 | - | 1 | 214 |