Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012755.1 | NP_036887.1 | 9 | 987 | ||
XM_006256520.2 | XP_006256582.1 | 7 | 1822 | |||
XM_006256522.1 | XP_006256584.1 | 8 | 1274 | |||
GenBank mRNA | U35365.1 | - | 9 | 987 |