Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012761.1 | NP_036893.1 | 3 | 1567 | ||
GenBank mRNA | U07610.1 | - | 3 | 1567 |