Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031551.1 | NP_113739.1 | 4 | 266 | ||
XM_006229551.1 | XP_006229613.1 | 2 | 688 | |||
XM_006229552.2 | XP_006229614.1 | 3 | 271 | |||
XM_006229553.2 | XP_006229615.1 | 4 | 291 | |||
XM_006229554.1 | XP_006229616.1 | 3 | 218 | |||
GenBank mRNA | BC083688.1 | - | 4 | 266 | 7189278 | |
FQ213717.1 | - | 4 | 145 | |||
FQ214482.1 | - | 4 | 232 | |||
FQ214526.1 | - | 4 | 232 | |||
FQ214664.1 | - | 4 | 207 | |||
FQ214718.1 | - | 4 | 282 | |||
FQ214896.1 | - | 4 | 217 | |||
FQ216691.1 | - | 4 | 222 | |||
FQ226989.1 | - | 3 | 144 | |||
X74125.1 | - | 4 | 192 |