Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031551.1 | NP_113739.1 | 13 | 1205 | ||
XM_006229551.1 | XP_006229613.1 | 11 | 1627 | |||
XM_006229552.2 | XP_006229614.1 | 12 | 1210 | |||
XM_006229553.2 | XP_006229615.1 | 13 | 1230 | |||
XM_006229554.1 | XP_006229616.1 | 12 | 1157 | |||
GenBank mRNA | BC083688.1 | - | 13 | 1205 | 7189278 | |
FQ213717.1 | - | 13 | 1084 | |||
FQ214526.1 | - | 13 | 1171 | |||
FQ214664.1 | - | 13 | 1146 | |||
FQ214718.1 | - | 13 | 1222 | |||
FQ214896.1 | - | 13 | 1155 | |||
FQ216691.1 | - | 13 | 1161 | |||
FQ217943.1 | - | 3 | 232 | |||
FQ226989.1 | - | 12 | 1082 | |||
X74125.1 | - | 13 | 1131 |