Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012780.1 | NP_036912.1 | 6 | 317 | ||
XM_006232850.2 | XP_006232912.1 | 6 | 444 | |||
XM_006232852.2 | XP_006232914.1 | 6 | 534 | |||
XM_006232853.2 | XP_006232915.1 | 5 | 399 | |||
XM_008761280.1 | XP_008759502.1 | 6 | 444 | |||
XM_008761281.1 | XP_008759503.1 | 5 | 399 | |||
GenBank mRNA | AY264361.1 | - | 6 | 317 | ||
AY264362.1 | - | 5 | 272 | |||
AY264363.1 | - | 6 | 317 | |||
AY264364.1 | - | 6 | 317 | |||
AY264365.1 | - | 6 | 317 | |||
U08986.1 | - | 1 | 24 | |||
U61184.1 | - | 6 | 317 |