Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012781.3 | NP_036913.3 | 14 | 1586 | ||
XM_006229494.2 | XP_006229556.1 | 15 | 2302 | |||
XM_006229495.2 | XP_006229557.1 | 14 | 2395 | |||
XM_006229496.2 | XP_006229558.1 | 14 | 2856 | |||
XM_006229497.2 | XP_006229559.1 | 14 | 2239 | |||
XM_006229498.2 | XP_006229560.1 | 14 | 1537 | |||
XM_008759602.1 | XP_008757824.1 | 14 | 1955 | |||
XM_008759603.1 | XP_008757825.1 | 14 | 1944 | |||
XM_008759604.1 | XP_008757826.1 | 14 | 4675 | |||
XM_008759605.1 | XP_008757827.1 | 14 | 1869 | |||
XM_008759606.1 | XP_008757828.1 | 14 | 2094 | |||
XM_008759607.1 | XP_008757829.1 | 14 | 1651 | |||
XM_008759608.1 | XP_008757830.1 | 13 | 1751 | |||
GenBank mRNA | AY264366.1 | - | 14 | 1515 | ||
DQ223732.1 | - | 1586 | ||||
U61157.1 | - | 14 | 1610 | |||
U61405.1 | - | 14 | 1586 |