Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012787.1 | NP_036919.1 | 7 | 1122 | ||
GenBank mRNA | M33296.2 | - | 7 | 1120 |