Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767172.1 | XP_008765394.1 | 18 | 3419 | ||
GenBank mRNA | D26498.1 | - | 4 | 354 |