Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012345.1 | NP_001012345.1 | 1 | 247 | ||
GenBank mRNA | BC089846.1 | - | 1 | 247 | 7372077 |