Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145780.1 | NP_665723.1 | 11 | 1484 | ||
XM_008765216.1 | XP_008763438.1 | 11 | 2131 | |||
XM_008765217.1 | XP_008763439.1 | 12 | 1848 | |||
GenBank mRNA | BC061822.1 | - | 12 | 1565 | 5599041 | |
FQ228928.1 | - | 6 | 764 | |||
L26398.1 | - | 11 | 1484 |