Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145782.1 | NP_665725.1 | 1, 2 | 118 | ||
GenBank mRNA | D38381.1 | - | 1, 2 | 118 | ||
X79991.1 | - | 1, 2 | 118 |