Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012828.2 | NP_036960.1 | 3, 4 | 378 | ||
XM_006257329.2 | XP_006257391.1 | 3, 4 | 450 | |||
XM_006257330.2 | XP_006257392.1 | 3, 4 | 859 | |||
GenBank mRNA | M88751.1 | - | 3, 4 | 382 |