Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203493.3 | NP_987089.2 | 1 | 23 | ||
XM_006250622.2 | XP_006250684.1 | 5 | 2596 | |||
XM_008769994.1 | XP_008768216.1 | 1 | 294 | |||
XM_008769995.1 | XP_008768217.1 | 4 | 713 | |||
GenBank mRNA | BC129082.1 | - | 1 | 23 | 8365946 | |
L11354.1 | - | 2 | 157 |