Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233067.2 | XP_006233129.1 | 2 | 324 | ||
XM_006233068.2 | XP_006233130.1 | 2 | 343 | |||
GenBank mRNA | DQ075463.1 | - | 2 | 655 | ||
M94152.1 | - | 970 | ||||
X59249.1 | - | 2 | 783 | |||
X93219.1 | - | 2 | 706 |
Gene Symbol: LOC100363178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749374.3 | XP_003749422.1 | 1474 | |||
XM_006233108.2 | XP_006233170.1 | 324 | ||||
XM_006233109.2 | XP_006233171.1 | 343 |
Set Membership: |
G-protein coupled receptor |