Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012949.2 | NP_037081.2 | 9, 10 | 1126 | ||
XM_006246599.2 | XP_006246661.1 | 9, 10 | 1197 | |||
XM_006246600.2 | XP_006246662.1 | 9, 10 | 1177 | |||
GenBank mRNA | BC083566.1 | - | 9, 10 | 1113 | 7193009 | |
FQ214810.1 | - | 8, 9 | 1105 | |||
FQ214945.1 | - | 9, 10 | 1144 | |||
FQ214984.1 | - | 9, 10 | 1138 | |||
FQ215197.1 | - | 5, 6 | 803 | |||
FQ215221.1 | - | 8, 9 | 1038 | |||
FQ215310.1 | - | 5, 6 | 804 | |||
FQ215357.1 | - | 9, 10 | 1149 | |||
FQ215526.1 | - | 9, 10 | 1137 | |||
FQ215648.1 | - | 9, 10 | 1136 | |||
FQ215759.1 | - | 9, 10 | 1148 | |||
FQ215780.1 | - | 5, 6 | 804 | |||
FQ215843.1 | - | 8, 9 | 1104 | |||
FQ215865.1 | - | 9, 10 | 1136 | |||
FQ216150.1 | - | 8, 9 | 1105 | |||
FQ216271.1 | - | 5, 6 | 804 | |||
FQ216382.1 | - | 9, 10 | 1139 | |||
FQ216386.1 | - | 8, 9 | 1114 | |||
FQ216781.1 | - | 5, 6 | 803 | |||
FQ217095.1 | - | 9, 10 | 1136 | |||
FQ223831.1 | - | 5, 6 | 803 | |||
FQ224938.1 | - | 9, 10 | 1136 | |||
Y00979.1 | - | 8, 9 | 1088 |