Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053296.1 | NP_445748.1 | 8, 9 | 1204 | ||
XM_006232506.2 | XP_006232568.1 | 9, 10 | 1350 | |||
GenBank mRNA | AJ310839.1 | - | 8, 9 | 1204 | ||
FQ212388.1 | - | 9, 10 | 1355 |