Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012981.2 | NP_037113.2 | 4 | 538 | ||
GenBank mRNA | BC085752.1 | - | 4 | 538 | 7130510 | |
D89863.1 | - | 946 |