Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013001.2 | NP_037133.1 | 3, 4 | 173 | ||
XM_006234633.2 | XP_006234695.1 | 4, 5 | 556 | |||
XM_006234634.2 | XP_006234696.1 | 4, 5 | 2012 | |||
XM_006234635.2 | XP_006234697.1 | 4, 5 | 471 | |||
XM_006234636.2 | XP_006234698.1 | 4, 5 | 1462 | |||
XM_008762060.1 | XP_008760282.1 | 3, 4 | 655 | |||
GenBank mRNA | AJ627631.1 | - | 13 | |||
AY540905.1 | - | 1, 2 | 28 | |||
AY540906.1 | - | 1, 2 | 28 | |||
BC128741.1 | - | 3, 4 | 173 | 8363963 | ||
HQ121510.1 | - | 14 | ||||
U69644.1 | - | 3, 4 | 393 |