Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013032.3 | NP_037164.3 | 12 | 1443 | ||
GenBank mRNA | BC061743.1 | - | 12 | 1443 | 5599027 | |
D63772.1 | - | 1474 | ||||
U21104.1 | - | 12 | 1455 | |||
U39555.1 | - | 12 | 1533 | |||
X94255.1 | - | 12 | 1500 |