Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749833.3 | XP_003749881.1 | 3, 4 | 610 |
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017102.2 | NP_058798.2 | 276 | |||
XM_006237295.1 | XP_006237357.1 | 504 | ||||
XM_006237296.2 | XP_006237358.1 | 823 | ||||
XM_006237297.1 | XP_006237359.1 | 508 | ||||
XM_006237298.2 | XP_006237360.1 | 786 | ||||
GenBank mRNA | BC063168.1 | - | 3, 4 | 403 | 6887820 | |
D13962.1 | - | 360 | ||||
U17978.1 | - | 276 |