Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013039.2 | NP_037171.2 | 25 | 2945 | ||
XM_008759319.1 | XP_008757541.1 | 24 | 3333 | |||
XM_008759320.1 | XP_008757542.1 | 25 | 3014 | |||
XM_008759321.1 | XP_008757543.1 | 25 | 2939 | |||
XM_008759322.1 | XP_008757544.1 | 25 | 3044 | |||
GenBank mRNA | AB052294.1 | - | 25 | 2960 | ||
AF039595.1 | - | 25 | 2945 | |||
AJ511272.1 | - | 25 | 2867 | |||
AJ511273.1 | - | 25 | 2924 | |||
X97279.1 | - | 25 | 2945 |