Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013063.2 | NP_037195.1 | 4 | 537 | ||
GenBank mRNA | BC085765.1 | - | 4 | 537 | 7107130 | |
U94340.1 | - | 4 | 505 |