Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013146.2 | NP_037278.1 | 10, 11 | 1668 | ||
XM_006236260.2 | XP_006236322.1 | 10, 11 | 3218 | |||
XM_006236261.2 | XP_006236323.1 | 11, 12 | 1739 | |||
XM_006236262.2 | XP_006236324.1 | 11, 12 | 1892 | |||
XM_008762763.1 | XP_008760985.1 | 12, 13 | 1820 |