Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013162.1 | NP_037294.1 | 5, 6 | 628 | ||
GenBank mRNA | BC167099.1 | - | 5, 6 | 629 | 7369314 | |
DV725071.1 | - | 3, 4 | 427 | |||
FQ218638.1 | - | 5, 6 | 644 | |||
M10934.1 | - | 4, 5 | 491 |