Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052807.2 | NP_434694.1 | 2 | 414 | ||
XM_006229324.2 | XP_006229386.1 | 2 | 414 | |||
XM_008759500.1 | XP_008757722.1 | 2 | 412 | |||
GenBank mRNA | L29232.1 | - | 2 | 414 | ||
M27293.1 | - | 2 | 414 |