Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022381.3 | NP_071776.1 | 4 | 590 | ||
GenBank mRNA | BC060570.1 | - | 4 | 675 | 6891516 | |
FQ219845.1 | - | 4 | 671 | |||
FQ219897.1 | - | 4 | 671 | |||
FQ220101.1 | - | 4 | 671 | |||
FQ220309.1 | - | 4 | 671 | |||
FQ220835.1 | - | 4 | 671 | |||
FQ220924.1 | - | 4 | 670 | |||
FQ225495.1 | - | 4 | 671 | |||
FQ225620.1 | - | 3 | 295 | |||
FQ226677.1 | - | 4 | 671 | |||
FQ226726.1 | - | 4 | 672 | |||
FQ227500.1 | - | 4 | 671 | |||
FQ227521.1 | - | 4 | 671 | |||
FQ229469.1 | - | 4 | 671 | |||
FQ229972.1 | - | 4 | 671 | |||
FQ230190.1 | - | 4 | 671 | |||
FQ232129.1 | - | 4 | 684 | |||
FQ233125.1 | - | 4 | 671 | |||
FQ233579.1 | - | 4 | 671 | |||
FQ233966.1 | - | 4 | 671 | |||
FQ234183.1 | - | 4 | 673 | |||
FQ234500.1 | - | 4 | 671 | |||
FQ234529.1 | - | 4 | 671 | |||
FQ234547.1 | - | 4 | 671 | |||
FQ234986.1 | - | 4 | 671 | |||
FQ235296.1 | - | 4 | 671 | |||
Y00047.1 | - | 4 | 590 |