Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013199.1 | NP_037331.1 | 19 | 2190 | ||
XM_006242604.2 | XP_006242666.1 | 20 | 2285 | |||
XM_006242605.2 | XP_006242667.1 | 20 | 2284 | |||
XM_006242608.2 | XP_006242670.1 | 19 | 2254 | |||
XM_006242610.2 | XP_006242672.1 | 19 | 2254 | |||
XM_006242611.2 | XP_006242673.1 | 19 | 2254 | |||
XM_006242612.2 | XP_006242674.1 | 18 | 2242 | |||
XM_006242613.2 | XP_006242675.1 | 18 | 2242 | |||
XM_006242614.2 | XP_006242676.1 | 19 | 2254 | |||
XM_008765917.1 | XP_008764139.1 | 20 | 2285 | |||
XM_008765918.1 | XP_008764140.1 | 19 | 2272 | |||
XM_008765919.1 | XP_008764141.1 | 20 | 2284 | |||
GenBank mRNA | L24562.1 | - | 19 | 2129 | ||
L25605.1 | - | 19 | 2190 |