Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013200.1 | NP_037332.1 | 14 | 1664 | ||
XM_006242180.2 | XP_006242242.1 | 14 | 2120 | |||
GenBank mRNA | BC085761.1 | - | 14 | 1676 | 7193798 | |
D43623.1 | - | 13 | 1651 |