Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195199.1 | NP_001182128.1 | 29 | 4177 | ||
NM_147141.1 | NP_671482.1 | 28 | 4205 | |||
XM_006233569.2 | XP_006233631.1 | 30 | 4323 | |||
XM_006233570.2 | XP_006233632.1 | 30 | 4320 | |||
XM_006233572.2 | XP_006233634.1 | 30 | 4323 | |||
XM_006233573.2 | XP_006233635.1 | 29 | 4311 | |||
XM_006233574.2 | XP_006233636.1 | 29 | 4311 | |||
XM_006233575.2 | XP_006233637.1 | 29 | 4260 | |||
XM_006233576.2 | XP_006233638.1 | 29 | 4257 | |||
XM_006233577.2 | XP_006233639.1 | 29 | 4260 | |||
XM_006233578.2 | XP_006233640.1 | 29 | 4257 | |||
XM_008761578.1 | XP_008759800.1 | 30 | 4323 | |||
GenBank mRNA | AF055477.2 | - | 28 | 4205 | ||
HQ008360.1 | - | 29 | 4177 | |||
M92905.1 | - | 29 | 4129 |