Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153468.1 | NP_703198.1 | 4, 5 | 617 | ||
GenBank mRNA | AB082125.1 | - | 4, 5 | 617 |