Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270871.1 | NP_001257800.1 | 25 | 2793 | ||
NM_001270872.1 | NP_001257801.1 | 24 | 2757 | |||
NM_181366.2 | NP_852031.1 | 26 | 2841 | |||
XM_006256916.2 | XP_006256978.1 | 26 | 2881 | |||
XM_008773154.1 | XP_008771376.1 | 25 | 3221 | |||
XM_008773155.1 | XP_008771377.1 | 25 | 2831 | |||
XM_008773156.1 | XP_008771378.1 | 24 | 3185 | |||
XM_008773157.1 | XP_008771379.1 | 24 | 3179 | |||
XM_008773158.1 | XP_008771380.1 | 23 | 3143 | |||
XM_008773159.1 | XP_008771381.1 | 22 | 2734 | |||
GenBank mRNA | AF538953.1 | - | 26 | 2851 | ||
AF538958.1 | - | 25 | 2803 | |||
AF538959.1 | - | 24 | 2767 |
Set Membership: |
G-protein coupled receptor |