Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172021.2 | NP_742018.2 | 9, 10 | 1831 | ||
XM_006247266.2 | XP_006247328.1 | 8, 9 | 2978 | |||
XM_006247267.2 | XP_006247329.1 | 6, 7 | 2119 | |||
XM_008767953.1 | XP_008766175.1 | 8, 9 | 2981 | |||
GenBank mRNA | AJ278800.1 | - | 8, 9 | 1817 | ||
BC076503.1 | - | 9, 10 | 1831 | 7100584 |