Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005761.1 | NP_001005761.1 | 8 | 1516 | ||
GenBank mRNA | AF526269.1 | - | 8 | 1125 | ||
BC083644.1 | - | 8 | 1516 | 7134963 |