Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004068.1 | NP_001004068.1 | 3 | 658 | ||
XM_006246969.2 | XP_006247031.1 | 3 | 732 | |||
XM_006246970.2 | XP_006247032.1 | 3 | 729 | |||
XM_006246973.2 | XP_006247035.1 | 3 | 969 | |||
XM_006246974.2 | XP_006247036.1 | 3 | 559 | |||
XM_006246975.2 | XP_006247037.2 | 4 | 1540 | |||
XM_006246977.2 | XP_006247039.1 | 3 | 1448 | |||
XM_006246978.2 | XP_006247040.1 | 3 | 966 | |||
XM_006246979.2 | XP_006247041.1 | 3 | 555 | |||
XM_008767954.1 | XP_008766176.1 | 4 | 1537 | |||
XM_008767955.1 | XP_008766177.1 | 4 | 1722 | |||
XM_008767956.1 | XP_008766178.1 | 4 | 1541 | |||
GenBank mRNA | AY157969.1 | - | 2 | 351 | ||
BC079216.1 | - | 3 | 658 | 7131890 | ||
FQ210900.1 | - | 3 | 440 |