Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173118.2 | NP_775141.2 | 4 | 483 | ||
GenBank mRNA | AJ515237.1 | - | 4 | 418 | ||
BC058132.1 | - | 4 | 446 | 6920130 | ||
FQ214040.1 | - | 4 | 404 | |||
FQ216545.1 | - | 4 | 408 | |||
FQ219301.1 | - | 4 | 425 | |||
FQ219863.1 | - | 4 | 416 | |||
FQ220660.1 | - | 4 | 404 | |||
FQ220919.1 | - | 4 | 410 | |||
FQ228456.1 | - | 4 | 408 | |||
FQ229422.1 | - | 4 | 463 | |||
FQ229782.1 | - | 4 | 404 | |||
FQ229783.1 | - | 4 | 406 | |||
FQ230107.1 | - | 4 | 410 |