Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173151.1 | NP_775174.1 | 7 | 746 | ||
XM_006257016.2 | XP_006257078.1 | 7 | 707 | |||
XM_006257017.2 | XP_006257079.1 | 7 | 1114 | |||
XM_008773254.1 | XP_008771476.1 | 7 | 827 | |||
GenBank mRNA | AF190256.1 | - | 7 | 746 |