Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105763.1 | NP_001099233.1 | 6 | 1621 | ||
XM_006245830.2 | XP_006245892.1 | 6 | 6293 | |||
XM_006245831.2 | XP_006245893.1 | 4 | 6037 |