Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cramp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246051.2 | XP_006246113.1 | 10 | 2462 | ||
XM_006246052.2 | XP_006246114.1 | 10 | 2126 | |||
XM_006246053.2 | XP_006246115.1 | 10 | 1912 | |||
XM_008767606.1 | XP_008765828.1 | 10 | 2462 |