Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246078.2 | XP_006246140.1 | 1, 2 | 208 | ||
XM_006246079.2 | XP_006246141.1 | 1, 2 | 226 | |||
XM_008767620.1 | XP_008765842.1 | 1, 2 | 208 |