Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109632.1 | NP_001103102.1 | 5 | 392 | ||
XM_006246402.2 | XP_006246464.1 | 4 | 510 |