Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013857.1 | NP_001013879.1 | 4, 5 | 578 | ||
XM_006246462.2 | XP_006246524.1 | 5, 6 | 846 | |||
XM_006246463.2 | XP_006246525.1 | 5, 6 | 625 | |||
XM_006246464.2 | XP_006246526.1 | 5, 6 | 1177 | |||
XM_006246465.2 | XP_006246527.1 | 5, 6 | 738 | |||
XM_008767782.1 | XP_008766004.1 | 6, 7 | 732 | |||
XM_008767783.1 | XP_008766005.1 | 5, 6 | 1942 | |||
XM_008767784.1 | XP_008766006.1 | 6, 7 | 2218 | |||
XM_008767785.1 | XP_008766007.1 | 5, 6 | 625 | |||
GenBank mRNA | BC083838.1 | - | 4, 5 | 578 | 7113397 |