Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105789.1 | NP_001099259.1 | 8 | 1647 | ||
XM_008767788.1 | XP_008766010.1 | 7 | 3094 | |||
XM_008767789.1 | XP_008766011.1 | 7 | 3654 | |||
XM_008767790.1 | XP_008766012.1 | 6 | 1983 | |||
XM_008767791.1 | XP_008766013.1 | 7 | 1831 | |||
XM_008767792.1 | XP_008766014.1 | 7 | 1829 | |||
XM_008767793.1 | XP_008766015.1 | 6 | 1985 |