Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105803.2 | NP_001099273.2 | 2 | 616 | ||
GenBank mRNA | CB326294.1 | - | 2 | 638 | ||
CO570263.1 | - | 2 | 616 |