Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011893.1 | NP_001011893.1 | 6 | 620 | ||
XM_008767966.1 | XP_008766188.1 | 10 | 1800 | |||
GenBank mRNA | AY208293.1 | - | 6 | 620 | ||
AY208294.1 | - | 6 | 620 | |||
BC088334.1 | - | 5 | 468 | |||
BC126069.1 | - | 6 | 521 | 7441949 |