Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011893.1 | NP_001011893.1 | 11 | 1150 | ||
XM_008767966.1 | XP_008766188.1 | 15 | 2383 | |||
GenBank mRNA | AY208293.1 | - | 11 | 1203 | ||
AY208294.1 | - | 11 | 1150 | |||
BC088334.1 | - | 10 | 1051 | |||
BC126069.1 | - | 10 | 1241 | 7441949 |