Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM187A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013861.1 | NP_001013883.1 | 1 | 554 | ||
GenBank mRNA | BC079299.1 | - | 1 | 554 | 7134519 |