Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009626.1 | NP_001009626.1 | 6, 7 | 789 | ||
GenBank mRNA | AM071386.1 | - | 6, 7 | 748 | ||
BC088180.1 | - | 6, 7 | 789 | 7364928 | ||
FQ209412.1 | - | 6, 7 | 828 | |||
FQ209491.1 | - | 6, 7 | 815 | |||
FQ209740.1 | - | 6, 7 | 825 | |||
FQ210311.1 | - | 6, 7 | 815 | |||
FQ210813.1 | - | 6, 7 | 815 | |||
FQ211328.1 | - | 6, 7 | 788 | |||
FQ218068.1 | - | 1, 2 | 102 | |||
FQ218335.1 | - | 2, 3 | 322 | |||
FQ218445.1 | - | 6, 7 | 829 | |||
FQ218454.1 | - | 6, 7 | 816 | |||
FQ218597.1 | - | 6, 7 | 811 | |||
FQ218599.1 | - | 6, 7 | 815 | |||
FQ218637.1 | - | 6, 7 | 815 | |||
FQ218785.1 | - | 6, 7 | 832 | |||
FQ218859.1 | - | 6, 7 | 788 | |||
FQ218865.1 | - | 6, 7 | 815 | |||
FQ218961.1 | - | 6, 7 | 815 | |||
FQ219138.1 | - | 6, 7 | 880 | |||
FQ219328.1 | - | 6, 7 | 815 | |||
FQ219380.1 | - | 6, 7 | 816 | |||
FQ219438.1 | - | 6, 7 | 814 | |||
FQ219481.1 | - | 6, 7 | 827 | |||
FQ219537.1 | - | 6, 7 | 812 | |||
FQ219580.1 | - | 6, 7 | 829 | |||
FQ219601.1 | - | 6, 7 | 828 | |||
FQ219674.1 | - | 6, 7 | 815 | |||
X15551.1 | - | 6, 7 | 644 |