Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105849.1 | NP_001099319.1 | 11 | 1138 | ||
XM_008768317.1 | XP_008766539.1 | 11 | 1172 | |||
XM_008768318.1 | XP_008766540.1 | 11 | 1310 | |||
XM_008768319.1 | XP_008766541.1 | 11 | 1172 | |||
XM_008768320.1 | XP_008766542.1 | 11 | 1252 | |||
XM_008768321.1 | XP_008766543.1 | 11 | 1746 |